Anti-Cleaved Caspase-7 Rabbit Monoclonal Antibody [clone: EPR22840-25]

Supplier: Abcam
AB276033-100UG AB276033-1MG
ABCA_AB276033-100UEA 1564.2 CAD
ABCA_AB276033-100U ABCA_AB276033-1MG
Anti-Cleaved Caspase-7 Rabbit Monoclonal Antibody [clone: EPR22840-25]
Antibodies
Rabbit monoclonal [EPR22840-25] to Cleaved Caspase-7 - BSA and Azide free (Capture).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cleaved Caspase-7
Clonality: Monoclonal
Clone: EPR22840-25
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR