Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E61] (Alexa Fluor® 488)

Supplier: Abcam
AB307610-100UL AB307610-10UL
ABCA_AB307610-100UEA 1248.2 CAD
ABCA_AB307610-100U ABCA_AB307610-10UL
Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E61] (Alexa Fluor® 488)
Antibodies
Alexa Fluor® 488 Rabbit monoclonal [E61] to pro Caspase-3.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: pro Caspase-3
Clonality: Monoclonal
Clone: E61
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR