Anti-CISH/CIS Rabbit Antibody [clone: EPR25357-36]

Supplier: Abcam
AB308500-1ML AB308500-100UL AB308500-20UL
ABCA_AB308500-1MLEA 11287.52 CAD
ABCA_AB308500-1ML ABCA_AB308500-100U ABCA_AB308500-20UL
Anti-CISH/CIS Rabbit Antibody [clone: EPR25357-36]
Antibodies
Rabbit monoclonal [EPR25357-36] to CISH/CIS.

Type: Primary
Antigen: CISH/CIS
Clonality:
Clone: EPR25357-36
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR