Anti-Topoisomerase II alpha+Topoisomerase II beta Rabbit Monoclonal Antibody [clone: EPR5377]

Supplier: Abcam
AB109524-100UL AB109524-1ML
ABCA_AB109524-100UEA 1049.12 CAD
ABCA_AB109524-100U ABCA_AB109524-1ML
Anti-Topoisomerase II alpha+Topoisomerase II beta Rabbit Monoclonal Antibody [clone: EPR5377]
Antibodies
Rabbit monoclonal [EPR5377] to Topoisomerase II alpha + Topoisomerase II beta/TOP2B.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Topoisomerase II alpha+Topoisomerase II beta
Clonality: Monoclonal
Clone: EPR5377
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR