Paragon Scientific Refractive Index (Nominal: 1.4196 @ 25 °C), Certified @ 20, 25 and 30 °C
ALK-PS-RI-04 ALK-PS-RI-04K
77975-906EA 216.46 CAD
77975-906 77975-907
Paragon Scientific Refractive Index (Nominal: 1.4196 @ 25 °C), Certified @ 20, 25 and 30 °C
Standards Refractive Index Standards
  • Standard type: Single component
  • Shipping conditions: Room temperature