Paragon Scientific Refractive Index (Nominal: 1.4941 @ 25 °C), Certified @ 20, 25 and 30 °C
ALK-PS-RI-07
ALK-PS-RI-07K
77975-910EA
216.46
CAD
77975-910
77975-911
Paragon Scientific Refractive Index (Nominal: 1.4941 @ 25 °C), Certified @ 20, 25 and 30 °C
Standards
Refractive Index Standards
Standard type: Single component
Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...