Paragon Scientific Density 25 °C (Nominal: 0.9342 g/ml @ 25 °C)
ALK-DEN25-06
ALK-DEN25-06-500ML
77975-689EA
153.26
CAD
77975-689
77975-690
Paragon Scientific Density 25 °C (Nominal: 0.9342 g/ml @ 25 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...