Paragon Scientific Density 15 °C (Nominal: 0.8648 g/ml @ 15 °C)
ALK-DEN15-06
ALK-DEN15-06-500ML
77975-648EA
153.26
CAD
77975-648
77975-649
Paragon Scientific Density 15 °C (Nominal: 0.8648 g/ml @ 15 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...