Paragon Scientific Density 25 °C (Nominal: 1.6136 g/ml @ 25 °C)
Supplier: LGC Standards
ALK-DEN25-13 ALK-DEN25-13-500ML
77975-700EA 153.26 CAD
77975-700 77975-701
Paragon Scientific Density 25 °C (Nominal: 1.6136 g/ml @ 25 °C)
Standards Density Standards
  • Shipping conditions: Room temperature