Paragon Scientific 5W30 Viscosity Check Oil (Kinematic Viscosity, CCS Dynamic Viscosity and Density)
ALK-CVCO5W30
ALK-CVCO5W30-5L
77975-587EA
93.22
CAD
77975-587
77975-588
Paragon Scientific 5W30 Viscosity Check Oil (Kinematic Viscosity, CCS Dynamic Viscosity and Density)
Standards
Viscometry Standards
- Standard type: Single component