PathFlow Supa Mega Base Mold
Supplier: StatLab
GBC-3614-05A GBC-6014-05A GBC-6014-05B
77908-056PK 231.14 CAD
77908-056 77908-058 77908-060
PathFlow Supa Mega Base Mold
Moulds Moulds for Histology
PathFlow stainless steel Supa Mega base molds.
  • Reusable
  • Stops wax leakage
  • Stackable design for efficient storage
  • Stainless steel for a lifetime of use