Trimming/Grossing Blades
Supplier: StatLab
GB-260MM-B GB-260MM-D GB-130MM-B GB-130MM-D
77908-052PK 699.89 CAD
77908-052 77908-054 77908-048 77908-050
Trimming/Grossing Blades
Scalpels and Scalpel Assemblies Scalpel Blades
Ultra-sharp, precision stainless steel blades in two lengths.
  • Blades are available in 130 and 260 mm lengths
Precisely honed steel allows for more precise trimming, uniform cuts, and consistent grossing on surgical/autopsy specimens. Available in user-friendly, safety dispenser packs, as well as in bulk format.