CellSafe+ Cassette Biopsy Capsule
Supplier: StatLab
EBE-0201-02A EBE-0301-02A
77908-040CS 182.28 CAD
77908-040 77908-042
CellSafe+ Cassette Biopsy Capsule
Embedding Cassettes Biopsy Processing Cassettes
Cellsafe+ is an easy-to-use alternative to the more traditional and time-consuming handling techniques such as tissue wrap and foam pads.
  • CellSafe+ is contained in a standard cassette for normal processing
  • Validated for use with standard cassettes
The CellSafe+ consists of two hinged interlocking frames with integral mesh. This extra fine x-ray transparent mesh ensures a high level of specimen security, zero tissue damage, and low reagent carry over. The capsule is closed in one direction to fully encapsulate the tissue. Prior to embedding, tissue can be removed with greater ease and security, compared with conventional foam pads or paper wrap.
The CellSafe+ biopsy capsule is a patented innovation by CellPath, a StatLab company.