Disposable Base Molds
Supplier: StatLab
BM1515 BM2424 BM3024 BM3724
77907-820CS 123.87 CAD
77907-820 77907-822 77907-824 77907-826
Disposable Base Molds
Moulds Moulds for Histology
Plastic base mold, approximately 5 mm height.
  • Clear plastic
  • Disposable design
  • 500 per case
Eliminates the use of cleaning solvents and release compounds.