StatPrep Adhesion CytoSlides
Supplier: VWR International
77907-996 77907-998
77907-996BX 90.28 CAD
77907-996 77907-998
StatPrep Adhesion CytoSlides
Slides Microscope Slides
StatPrep adhesion cytoslides are designed for use with the StatPrep liquid-based cytology collection system.
  • Small and large circle formats
  • Small circle: 13 mm
  • Large circle: 36 mm