Parafilm™ M Self-Sealing Flexible Film
Supplier: StatLab
PM992 PM996
77908-106RL 140.45 CAD
77908-106 77908-108
Parafilm™ M Self-Sealing Flexible Film
Seals General Purpose Sealing Films
Made of a semi-transparent, flexible, and water-proof material.
  • Available in 2 or 4" widths
Film is resistant to most acids and alcohols.