Anti-USP3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A72183-100UG A72183-50UG
77845-780EA 521.4 CAD
77845-780 77845-781
Anti-USP3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR