Anti-PECAM1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74399-100UL A74399-50UL
77849-558EA 521.4 CAD
77849-558 77849-559
Anti-PECAM1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: PECAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR