Anti-PFKFB2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73893-100UL A73893-50UL
77848-712EA 521.4 CAD
77848-712 77848-713
Anti-PFKFB2 Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: PFKFB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR