Anti-MDFIC Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A71054-100UL A71054-50UL
77843-663EA 521.4 CAD
77843-663 77843-664
Anti-MDFIC Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MDFIC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR