Anti-FBXO3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A71627-100UG A71627-50UG
77844-731EA 521.4 CAD
77844-731 77844-732
Anti-FBXO3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: FBXO3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR