Anti-DDX19A Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A70977-100UG A70977-50UG
77843-517EA 521.4 CAD
77843-517 77843-518
Anti-DDX19A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: DDX19A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR