Anti-HMGB1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74674-100UL A74674-50UL
77850-015EA 521.4 CAD
77850-015 77850-016
Anti-HMGB1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:2000, IHC:1:10 to 1:50

Type:
Antigen: HMGB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR