Human Growth Hormone (from Pituitary)
Supplier: MilliporeSigma
869008-100UG 869008-50UG
CA80055-048EA 1084.29 CAD
CA80055-048 CA80055-050
Human Growth Hormone (from Pituitary)
Proteins and Peptides
Growth hormone, Human pituitary, iodination grade, growth hormone, Human pituitary, iodination grade, CAS 12629-01-5, is a single chain polypeptide hormone essential for growth of all tissues and plays a role in fat mobilization.