Anti-MUSK Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74542-100UL A74542-50UL
77849-791EA 521.4 CAD
77849-791 77849-792
Anti-MUSK Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:5000, IHC: 1:50 to 1:200

Type:
Antigen: MUSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR