Anti-DDX11 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73968-100UL A73968-50UL
77848-832EA 521.4 CAD
77848-832 77848-833
Anti-DDX11 Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: DDX11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR