Anti-KCND3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73470-100UL A73470-50UL
77848-012EA 521.4 CAD
77848-012 77848-013
Anti-KCND3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000

Type:
Antigen: KCND3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR