Anti-MAP1A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A72843-100UL A72843-50UL
77846-973EA 521.4 CAD
77846-973 77846-974
Anti-MAP1A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR