Anti-HIST1H3A HEK293F cells Recombinant Monoclonal Antibody [clone: 2G10]

Supplier: AFG Bioscience
A74845-100UL A74845-50UL
77850-262EA 592.5 CAD
77850-262 77850-263
Anti-HIST1H3A HEK293F cells Recombinant Monoclonal Antibody [clone: 2G10]
Antibodies
Type:
Antigen: HIST1H3A
Clonality: Recombinant Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR