Anti-ITGB3BP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74359-100UL A74359-50UL
77849-491EA 521.4 CAD
77849-491 77849-492
Anti-ITGB3BP Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: ITGB3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR