Anti-CST11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A70741-100UG A70741-50UG
77843-074EA 521.4 CAD
77843-074 77843-075
Anti-CST11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: CST11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR