Anti-ZKSCAN5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A72353-100UG A72353-50UG
77846-100EA 521.4 CAD
77846-100 77846-101
Anti-ZKSCAN5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: ZKSCAN5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol ZKSCAN5
Environmentally Preferable
Antigen synonyms KIAA1015|ZFP95|Zinc finger protein with KRAB and SCAN domains 5|Zinc finger protein 95 homolog|Zfp-95
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9Y2L8
Isotype IgG
ELISA Yes
Immunogen Recombinant human Zinc finger protein with KRAB and SCAN domains 5 protein (490-643AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR