Anti-RAD54L2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A72241-100UG A72241-50UG
77845-889EA 521.4 CAD
77845-889 77845-890
Anti-RAD54L2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: RAD54L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol RAD54L2
Environmentally Preferable
Antigen synonyms Androgen receptor interacting protein 4|Androgen receptor-interacting protein 4|Arip4|ARIP4_HUMAN|HCMV interacting protein|Helicase ARIP4|HSPC325|RAD54 like 2 (S. cerevisiae)|RAD54 like protein 2|RAD54-like protein 2|SRISNF2L|Steroid receptor interacting SNF2 domain protein
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9Y4B4
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Helicase ARIP4 protein (1122-1296AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR