Anti-AIF1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74178-50UL A74178-100UL
77849-194EA 355.5 CAD
77849-194 77849-193
Anti-AIF1 Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR