Anti-POLI Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A71667-100UG A71667-50UG
77844-809EA 521.4 CAD
77844-809 77844-810
Anti-POLI Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: POLI
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR