Anti-PCK1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73218-100UL A73218-50UL
77847-592EA 521.4 CAD
77847-592 77847-593
Anti-PCK1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR