Anti-PADI2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A71963-100UG A71963-50UG
77845-362EA 521.4 CAD
77845-362 77845-363
Anti-PADI2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: PADI2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol PADI2
Environmentally Preferable
Antigen synonyms KIAA0994|OTTHUMP00000044625|PAD 2|PAD H19|PAD-H19|PAD2|PADI 2|PADI2 protein|PADI2_HUMAN|PDI 2|PDI2|Peptidlyarginine deiminase type II|Peptidyl arginine deiminase II|Peptidyl arginine deiminase type II|Peptidylarginine deiminase II|Protein arginine deiminase|Protein arginine deiminase type 2|Protein arginine deiminase type II|Protein-arginine deiminase type II|Protein-arginine deiminase type-2
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9Y2J8
Isotype IgG
ELISA Yes
Immunogen Recombinant human Protein-arginine deiminase type-2 protein (1-665AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR