Anti-HIST1H3A (mono/di/tri-methyl-K14) HEK293F cells Recombinant Monoclonal Antibody [clone: 1B2]

Supplier: AFG Bioscience
A74840-100UL A74840-50UL
77850-256EA 592.5 CAD
77850-256 77850-257
Anti-HIST1H3A (mono/di/tri-methyl-K14) HEK293F cells Recombinant Monoclonal Antibody [clone: 1B2]
Antibodies
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR