Anti-TENM1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A71729-100UL A71729-50UL
77844-927EA 521.4 CAD
77844-927 77844-928
Anti-TENM1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: TENM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR