Anti-TEKT5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73848-100UL A73848-50UL
77848-635EA 521.4 CAD
77848-635 77848-636
Anti-TEKT5 Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: TEKT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR