Anti-CORIN Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A72272-100UL A72272-50UL
77845-944EA 521.4 CAD
77845-944 77845-945
Anti-CORIN Rabbit Recombinant Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: CORIN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR