Anti-DDX43 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74626-100UL A74626-50UL
77849-935EA 521.4 CAD
77849-935 77849-936
Anti-DDX43 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:200 to 1:1000, IHC:1:50 to 1:200

Type:
Antigen: DDX43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR