Anti-MYH2 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A71839-100UL A71839-50UL
77845-131EA 521.4 CAD
77845-131 77845-132
Anti-MYH2 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: MYH2
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol MYH2
Environmentally Preferable
Antigen synonyms adult 2|Fast 2a myosin heavy chain|IBM3|Inclusion body myopathy 3, autosomal dominant|MYH2_HUMAN|MYH2A|MYHas8|MyHC IIa|MyHC-2a|MyHC-IIa|MYHSA2|Myosin heavy chain 2|Myosin heavy chain 2a|Myosin heavy chain|Myosin heavy chain IIa|Myosin heavy chain skeletal muscle adult 2|Myosin heavy polypeptide 2 skeletal muscle adult|Myosin-2|MYPOP|skeletal muscle|Type IIA myosin heavy chain
Clonality Recombinant Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID Q9UKX2
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Myosin-2 protein (1154-1300AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80°C, avoid repeated freeze
Shipping temperature Shipped on Blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR