Anti-HOXB13 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74223-100UL A74223-50UL
77849-267EA 521.4 CAD
77849-267 77849-268
Anti-HOXB13 Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: HOXB13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR