Anti-MKLN1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A71278-100UG A71278-50UG
77844-077EA 521.4 CAD
77844-077 77844-078
Anti-MKLN1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MKLN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR