Anti-GST Tag Mouse Monoclonal Antibody

Supplier: MilliporeSigma
71097-3 71097-4
CA80600-400EA 1158.68 CAD
CA80600-400 CA80600-402
Anti-GST Tag Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GST Tag
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR