Anti-MAFB Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A72613-100UG A72613-50UG
77846-586EA 521.4 CAD
77846-586 77846-587
Anti-MAFB Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000

Type:
Antigen: MAFB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR