Anti-ADCY5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A72698-100UL A72698-50UL
77846-737EA 521.4 CAD
77846-737 77846-738
Anti-ADCY5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:50 to 1:200

Type:
Antigen: ADCY5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR