Anti-H1F0 HEK293F cells Recombinant Monoclonal Antibody [clone: 1C7]

Supplier: AFG Bioscience
A74819-50UL A74819-100UL
77850-234EA 371.3 CAD
77850-234 77850-233
Anti-H1F0 HEK293F cells Recombinant Monoclonal Antibody [clone: 1C7]
Antibodies
Type:
Antigen: H1F0
Clonality: Recombinant Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR