Anti-HECW2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A71068-100UG A71068-50UG
77843-687EA 521.4 CAD
77843-687 77843-688
Anti-HECW2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: HECW2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol HECW2
Environmentally Preferable
Antigen synonyms C2 and WW domain-containing protein 2|E3 ubiquitin protein ligase E3 ubiquitin-protein ligase HECT|HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2|HECW2_HUMAN|KIAA130|KIAA1301|NEDD4 like E3 ubiquitin protein ligase 2|NEDD4 Related E3 Ubiquitin Ligase|NEDD4-like E3 ubiquitin-protein ligase 2
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9P2P5
Isotype IgG
ELISA Yes
Immunogen Recombinant human E3 ubiquitin-protein ligase HECW2 protein (495-641AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR